Would you like to log in again?
EnzyChrom™ Monoamine Oxidase Inhibitor Screening Kit, BioAssay Systems
Supplier: BioAssay Systems
EIMO-100
76236-130EA 552.8 USD InStock
76236-130
EnzyChrom™ Monoamine Oxidase Inhibitor Screening Kit, BioAssay Systems
Assays Enzyme Assays
HTS for inhibitor screening and evaluation of monoamine oxidase inhibitors.
  • Homogeneous and convenient
  • "Mix-incubate-measure" type assay
  • No wash and reagent transfer steps are involved
  • Robust and amenable to HTS: can be readily automated on HTS liquid handling systems for processing thousands of samples per day
  • Method: FL530/585 nm
  • Samples: Monoamine Oxidase
Monoamine oxidase (MAO, EC 1.4.3.4) are a family of mitochondrial enzymes that catalyze the oxidative deamination of monoamines. Two isoforms of MAO exist, MAO-A and MAO-B, with different inhibitor selectivity and tissue distribution. MAO dysfunction is thought to be responsible for a number of neurological disorders.
Unusually high or low levels of MAOs in the body have been associated with depression, schizophrenia, substance abuse, attention deficit disorder, migraines, and irregular sexual maturation. MAO inhibitors are one of the major classes of drug prescribed for the treatment of depression, Parkinson’s and Alzheimer's diseases. MAO Inhibitor Screening Assay Kit provides a convenient fluorimetric means to screen for MAO enzyme inhibitors. In the assay, MAO reacts with p-tyramine, a substrate for both MAO-A and MAO-B, resulting in the formation of H₂O₂, which is determined by a fluorimetric method (λem/ex = 585/530 nm). The assay is simple, sensitive, stable and high-throughput adaptable.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty