Would you like to log in again?
GenePrint® 10 System, Promega
Supplier: Promega
GenePrint®
B9510
76201-514EA 941.78 USD InStock
76201-514
GenePrint® 10 System, Promega
Nucleic Acid Reagents End-point PCR Enzymes and Kits
STR analysis for human cell line authentication.
  • Amplifies ANSI-0002-Recommended Loci
  • Tolerates higher DNA amounts
  • Better balance for aneuploid samples
  • Reduced PCR Time
To standardize STR analysis for human cell line authentication, the American Tissue Culture Collection (ATCC) Standards Development Organization Workgroup published the ASN-0002-2011, which recommends the use of at least eight STR loci (TH01, TPOX, vWA, CSF1PO, D16S539, D7S820, D13S317 and D5S818) plus Amelogenin for gender identification.
Delivery information: It includes GenePrint® 10 5X Primer Pair Mix - 250 μl, GenePrint® 10 5X Master Mix - 250 μl, GenePrint® 10 Allelic Ladder Mix - 50 μl, 2800M Control DNA - 25 μl (10 ng/μl), Internal Lane Standard 600 - 150 μl, Water Amplification Grade - 1250 μl.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty