Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HDAC6 Mouse Monoclonal Antibody [clone: 3D2]

Supplier: BioVision
AB253034-50UG
76265-994EA 781.42 USD InStock
76265-994
Anti-HDAC6 Mouse Monoclonal Antibody [clone: 3D2]
Antibodies
The affinity purified mouse monoclonal antibody is immunospecific for the D2 domain (a.a. 471-481) of human histone deacetylase 6 (HDAC6) as determined by ELISA and peptide library screening. HDAC6 belongs to the class II of the histone deacetylase family (EC 3.5.1.98). The protein of molecular mass 131 kDa consists of two catalytic domains, the serine-glutamate-rich SE14 domain and the ubiquitin-binding ZnF domain. HDAC6 shuttles between the nucleus and the cytoplasm, but its localization is predominantly cytoplasmic. The enzyme deacetylates various substrates including α-tubulin, HSP90, cortactin, histones and other proteins. Described protein plays an important role in microtubule-dependent cell motility, apoptosis, cell signaling and transcriptional regulation, protein degradation, immune response and DNA repair. HDAC6 is linked to several neuronal diseases and carcinogenesis

ELISA: 0.5 – 5µg/ml; Immunocytochemistry/Immunofluorescence: 5 – 50 µg/ml; WB: 0.5-5µg/ml

Type: Primary
Antigen: HDAC6
Clonality: Monoclonal
Clone: 3D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty