Would you like to log in again?
Human Recombinant PEPCK (from E. coli)
Supplier: BioVision
AB229508-10UG
76265-882EA 390.68 USD InStock
76265-882
Human Recombinant PEPCK (from E. coli)
Enzymes
  • Gene ID: 5105
  • Accession number: P35558
Phosphoenolpyruvate Carboxykinase (PCK1) belongs to the lyase family. In the presence of GTP, it catalyzes the reversible conversion of oxaloacetate (OAA) into phosphoenolpyruvate (PEP), GDP and CO2. In humans, two isoforms of PEPCK are found: cytosolic form (PCK1, also called PEPCK-C) and mitochondrial form (PEPCK-M). PCK1 is a control point enzyme which catalyzes a irreversible step in gluconeogenesis.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty