Would you like to log in again?
Rabbit Native Actin (from Muscle)
Supplier: BioVision
AB286054-1 AB286054-500UG
76265-914EA 219.64 USD InStock
76265-914 76265-912
Rabbit Native Actin (from Muscle)
Proteins and Peptides
Actin is one of the major components of cytoskeleton, highly conserved and abundant proteins in eukaryotic cells. It is present as monomeric globular protein, called G actin or as a polymer filament, F actin. It plays major role in cell division, cell motility, cell signaling, organelle movement etc. Mammals have 6 isoforms of Actin, which can be divided into 3 classes, α, β and γ. Muscle Actin is of α class and all other non muscle actin fall in β and γ -class.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty