Would you like to log in again?
Cells for Colorimeter, Type 93, Firefly
Supplier: FireFlySci
93G2 93G10 93G30 93G20 93G5 93G50
76016-364EA 551.49 USD InStock
76016-364 76016-354 76016-362 76016-358 76016-356 76016-360
Cells for Colorimeter, Type 93, Firefly
Cuvettes Spectrophotometer Cuvettes
The Type 93 cells are made from optical glass with a wavelength range of 340 to 2500 nm for use in colorimeters.
  • Lightpaths from 2 to 50 mm available
  • Excellent transmission values
  • Two windows optically polished
  • Fire-fused to ensure that cuvettes are robust and durable
  • Made by hand, ensuring optical polishing with no defects
Fits ACS, Data Color, Hunter and several other colorimeters. Each cell is vigorously inspected and delivers high accurate results.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty