Would you like to log in again?
Inland® 45 Mechanical Pump Oil, Scientific Instrument Services
Supplier: Scientific Instruments
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
INV451
MSPP-INV451EA 249.92 USD InStock
MSPP-INV451
Inland® 45 Mechanical Pump Oil, Scientific Instrument Services
Lubricants
Inland 45 is the rough pump oil suitable for mass spectrometer.
  • Non-toxic
  • Non-corrosive
  • Reclaimable
Compatible with Buna-N, neoprene and Viton elastomers and miscible with petroleum based oils, freon, aromatic solvents and flushing fluids.
It permits strain-free cold starts as a result of its low pour point and high viscosity index (its viscosity is lower than that of typical vacuum pump oils at low temperatures). It maintains its viscosity at high temperatures, it provides a good seal under full load conditions. It has unique molecular structure, chemical inertness and high-temperature stability superior to those of other high-performance oils, such as white oils and low vapor pressure (10-7 torr at 25 °C).
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty