Would you like to log in again?
Microtubes, Prepared, Sarstedt
Supplier: Sarstedt
41.1500.005
MSPP-411500005PK 85.27 USD InStock
MSPP-411500005
Microtubes, Prepared, Sarstedt
Tubes Microtubes
This translucent polypropylene tube is designed with attached brown color polyethylene push cap closure, serum gel with clotting activator.
  • Skirted, conical base
  • Transparent label with lot and shelf life
This tube can hold the sample volume of up to 1.1 ml.
Certifications: CE certified.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty