Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

STAT5 inhibitor ≥97% (by HPLC), Calbiochem®

Supplier: MilliporeSigma
Synonyms: N'-((4-Oxo-4H-chromen-3-yl)methylene)nicotinohydrazideNʹ-((4-Oxo-4H-chromen-3-yl)methylene)nicotinohydrazide

573108-10MG 573108-10MG
95062-676EA 151.56 USD InStock
CA95062-676 95062-676
STAT5 inhibitor ≥97% (by HPLC), Calbiochem®
STAT5 inhibitor
STAT5 inhibitor is a cell-permeable compound that selectively targets the SH2 domain of STAT5 (IC50 = 47 µM against STAT5b SH2 domain EPO peptide binding activity).


Formula: C₁₆H₁₁N₃O₃
MW: 293.282 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00124694
CAS Number: 285986-31-4
Order Now
Specification Test Results
Purity (HPLC) ≥97%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty