Would you like to log in again?
Multi-Fit Cap for Tubes, Sarstedt
Supplier: Sarstedt
65.1379
MSPP-651379CS 33.08 USD InStock
MSPP-651379
Multi-Fit Cap for Tubes, Sarstedt
Closures Test/Sample Tube Closures
Suitable for S-Monovette® of 13 mm diameter and tubes of 12 to 17 mm diameter.
  • Can be used in two different positions
  • Compact stack pack packing with round base
  • Suitable for deep freezing
  • Centrifugation upto 4,500×g, 10 mins
The conical false bottom especially suitable for small volumes and platform compatibility. Meets IATA requirements for sample support.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty