Would you like to log in again?
Butterfly Needle Winged Infusion Set, SAI Infusion Technologies
Supplier: SAI Infusion Technologies
BF-27-13
MSPP-BF2713EA 13.11 USD InStock
MSPP-BF2713
Butterfly Needle Winged Infusion Set, SAI Infusion Technologies
Animal Supplies Animal Infusion Supplies
These specialty needles (often called Winged Infusion Sets) offer a highly effective means of accessing a peripheral blood vessel for infusion or sampling in small and large animals.
  • Wings provide added grip and accuracy when handling the needle
  • Low-profile needle hub allows parallel needle insertion
  • Wide variety of needle sizes and tubing lengths available
  • Often used for rodent tail vein access or in the cephalic or saphenous veins of larger species
  • Connect to standard syringes or a luered fitting
  • Customizable
When compared to traditional straight needles, butterfly needles are often shorter with a low-profile hub, and include wings on both sides of the needle for stability. They have tubing attached to the end for easy connection to syringes or collection devices. Supplied sterile.
Packaging: Packaged individually and sterile.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty