Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glass Fiber Spore Discs, Crosstex®

Supplier: Crosstex
GFTS-06
75840-974PK 712.22 USD InStock
75840-974
Glass Fiber Spore Discs, Crosstex®
Sterilization Indicators
Glass Fiber Spore Discs for use in monitoring Plasma/Vaporized Hydrogen Peroxide (VH2O2) sterilization processes. These Biological Indicators are designed for monitoring processes whether placed directly into the chamber, positioned in areas of a device which are the most difficult to sterilize, or distributed throughout rooms and isolators.

  • Discs are 9mm in diameter
  • Shelf life of 24 months
  • Storage conditions of 5 to 30 °C, 20 to 70% relative humidity

A reduced incubation time of 24 hours when used with Geobacillus stearothermophilus have been validated when used in conjunction with prepared culture media.

Certifications: Certified with population, purity, and resistance characteristics (D value, survival and kill where applicable) and ISO 11138-1 compliant.

Delivery information: Certificate of Analysis included.

Packaging: Available in easy-to-access Tyvek®/Mylar® pouches.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty