Would you like to log in again?
Addista Mixed-Parameter Standard, Low Range, Hach
Supplier: Hach
Addista
LCA721
75921-226EA 109.34 USD InStock
75921-226
Addista Mixed-Parameter Standard, Low Range, Hach
Standards Environmental Analysis Standards
Mixed-Parameter quality control standard used for wastewater parameters.
  • Ready-to use
  • Less complexity
  • Less hands-on-time and eliminates an additional error source when analysing standard solutions
Applicable for the cuvette tests
- LCI500 COD (ISO 15705 formulation), 0 to 150 mg/L O₂
- APC500 COD (ISO 15705 formulation), 0 to 150 mg/L O₂
- APC314 COD, 15 to 150 mg/L O₂
- APC304 Ammonium, 0.015 to 2.0 mg/L NH₄-N
- APC138 Total Nitrogen, 5 to 40 mg/L TNb
- APC339 Nitrate, 0.23 to 13.5 mg/L NO₃-N
- APC349 Phosphate, 0.05 to 1.5 mg/L PO₄-P
Certifications: Components of the standard solution are traceable to SRM from NIST.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty