Would you like to log in again?
Marking Pens, Leica Biosystems
Supplier: Leica
3801880
103051-182PK 93.84 USD InStock
103051-182
Marking Pens, Leica Biosystems
Pens Permanent Markers
These marking pens are extremely versatile and can be used on various writing surfaces found in the laboratory.
  • Quick drying ink
  • Highly resistant to aqueous based stains, alcohols, xylene and xylene substitutes
  • Tight fitting cap helps prevent drying of the tip
  • Pen barrel is ergonomically designed for ease of use
They have an extra fine point made of rigid nylon for precise marking on glass slides as well as tissue cassettes.
Packaging: Packed in 1 box of 12 pieces.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty