Would you like to log in again?
Wash Bottles, Plastic, HCL Labels
Supplier: HCL Labels
SWB-0007-0016 SWB-0010-0016 SWB-0006-0016 SWB-0004-PL-0016 SWB0008PL-0016 BOT1839 SWB-0001-0016 SWB-0002-0016
103257-026EA 18.52 USD InStock
103257-026 103257-030 101417-521 103257-032 101417-525 101417-523 103257-028 101417-515
Wash Bottles, Plastic, HCL Labels
Bottles Wash Bottles
Empty wash bottles ensure leak-proof dispensing of common solvents, and are a great value.
  • Solvent-resistance print on bottles
  • Convenient as a simple 'fill and go' option
  • Not designed for storage of solvents, however they are solvent resistant
Empty bottles are pre-printed with DOT, NFPA codes, and CAS numbers. These protective symbols and target information will keep all workstations a safer place.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty