Would you like to log in again?
Spatula Set, United Scientific Supplies
Supplier: United Scientific Supplies
SPATST6-VWR
76263-358EA 25.19 USD InStock
76263-358
Spatula Set, United Scientific Supplies
Spatulas
Complete set of six spatulas for a variety of needs.
  • Wide range of sizes
  • Includes storage box
The most popular spoons and spatulas for the laboratory includes one each of the following: 4" blade mounted on a hardwood handle; micro spoon with plastic handle; stainless steel 6" spatula (one end flat, one end spoon); 6" stainless steel spatula (one end bent, one end flat); 6" stainless steel spatula (one end flat, one end tapered); and one polypropylene 6" spatula (one end spatula, one end spoon).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty