Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

GMS Staining Kit, Biovision

Supplier: BioVision
AB287884-125
76265-688EA 652.3 USD InStock
76265-688
GMS Staining Kit, Biovision
Stains Microbiology Stains
The GMS Stain Kit, (modified Gomori Methenamine-Silver Nitrate) is intended for use in the histologic visualization of fungi, basement membrane and some opportunistic organisms such as Pneumocystis carinii.

  • For in vitro diagnostic use

Kit Components
• Silver Nitrate Solution (0.2%)
• Methenamine Solution
• Gold Chloride Solution (0.2%)
• Borax Solution
• Sodium Bisulfite Solution
• Chromic Acid Solution
• Sodium Thiosulfate Solution (5%)
• Light Green Solution

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty