Would you like to log in again?
PCR Tube, Forensic DNA Grade, Eppendorf
Supplier: Eppendorf
0030124707
76019-010CS 125.93 USD InStock
76019-010
PCR Tube, Forensic DNA Grade, Eppendorf
Tubes PCR Tubes
The Forensic DNA Grade product line encompasses consumables for DNA extraction, sample processing and PCR setup as well as for sample storage.
  • Positive air pressure prevents intrusion of dust
  • Strict adherence to cleaning procedures and protocols minimize the risk of contamination
A high degree of automation to minimize direct contact between staff and product.
Certifications: Certified purity grade according to ISO 18385.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty