Would you like to log in again?
Fatty Acid Uptake Assay Kit (Cell-Based), BioVision Inc.
Supplier: BioVision
AB287857-100TEST
76265-734EA 715.49 USD InStock
76265-734
Fatty Acid Uptake Assay Kit (Cell-Based), BioVision Inc.
Assays Cellular Assays
Non-radioactive fluorometric assay to detect free fatty uptake in adherent cells.
  • Simple one step procedure
  • Fast and convenient
  • The assay is easy, non-radioactive and highly sensitive
Fatty acids are essential dietary nutrients consisting of a carboxylic acid moiety linked to a long aliphatic hydrocarbon chain. They are the fundamental building blocks of more complex lipids such as those that form cellular membranes. In the blood, long-chain fatty acids travel bound to serum albumin and are transported into cells by members of the SLC27 family of transmembrane transporter proteins.
Uptake of long-chain free fatty acids into adipocytes is essential for regulating circulating lipid levels and cellular lipid metabolism. Abnormalities in adipocyte fatty acid uptake and lipid metabolism are major contributing factors to obesity, diabetes/metabolic syndrome, cardiovascular disorders and nonalcoholic fatty liver disease (NAFLD).
As diet and exercise consistently fail to achieve any lasting results in combating obesity, pharmacological targeting of fatty acid uptake is being investigated as a potential treatment strategy. BioVision’s Fatty Acid Uptake Assay Kit enables rapid determination of real-time fatty acid uptake kinetics in live cells expressing fatty acid transporter proteins.
The assay uses a highly fluorescent long-chain fatty acid analogue that acts as a substrate for transmembrane fatty acid transporters (FATPs) and accumulates in intracellular lipid droplets. A proprietary non-toxic membrane-impermeable quenching agent eliminates any fluorescence arising from the extracellular space, ensuring specific measurement of intracellular fatty acid accumulation without requiring any tedious wash steps.
The assay is highly sensitive, has a simple ‘mix-and-read’ protocol and is high-throughput adaptable for screening of fatty acid uptake-modulating test compounds or treatment conditions.
Delivery information: The kit contains a complete set of reagents sufficient for performing 100 reactions in a 96-well plate format.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty