Would you like to log in again?
PPDK Activity Assay Kit (Fluorometric/Colorimetric), BioVision Inc.
Supplier: BioVision
AB273318-100TEST
76265-738EA 819.71 USD InStock
76265-738
PPDK Activity Assay Kit (Fluorometric/Colorimetric), BioVision Inc.
Assays Enzyme Assays
Assay for measuring PPDK (Pyruvate, phosphate dikinase) in various bio samples such as bacterial homogenates, plant tissues.
  • Simple procedure takes less than 60 minutes
  • Fast and convenient
Pyruvate, phosphate dikinase, PPDK, (E.C. 2.7.9.1) is found in microorganisms and plants where it catalyzes reversible interconversion of AMP, PPi and phosphoenolpyruvate (PEP) to ATP, Pi and pyruvate monophosphate.
PPDK plays an important role in plant photosynthesis and glycolytic pathway of endobiotic protozoa and bacteria where pyruvate, phosphate dikinase (PK) is absent, it functions in the process of ATP biosynthesis. In plants, PPDK is a pivotal regulator of the C4 photosynthetic cycle and the key limiting factor for maintaining photosynthesis at low temperature.
It catalyzes the regeneration of PEP as the primary acceptor of atmospheric CO2 from pyruvate, ATP and phosphate. In addition, PPDK is also present in C3 leaves where it confers a similar light/dark regulation in chloroplasts, and in wheat leaves in an amount comparable to that found in seeds. Since PPDK is not found in vertebrates, finding its inhibitors may be useful for treatment of infections caused by anaerobic protists that depend on pyrophosphate-dependent glycolysis.
Specific inhibition of PPDK is expected to inhibit Giardia and other anaerobic protists such as Entamoeba histolytica and Trichomonas vaginalis without harming their mammalian hosts. This Pyruvate, phosphate dikinase, (PPDK) Assay Kit provides a simple and rapid test for measuring specific activity of PPDK in bacterial, protozoan and plant lysates as well as protein preps. This utilize the ability of PPDK to interconvert the substrate into intermediate product detected by proprietary enzyme mix and probe.
Generated stable product can be quantified by either colorimetric or fluorometric readout. The assay is simple to perform, high-throughput adaptable and a fluorometric reaction can detect as low as of 0.006 U of PPDK activity in a single well. We provide sufficient reagents for 100 fluorometric or colorimetric assays.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty