Would you like to log in again?
EZClick™ Global Protein Synthesis Assay Kit (FACS/Microscopy), Green Fluorescence, BioVision Inc.
Supplier: BioVision
AB239725-100TEST
76265-766EA 913.27 USD InStock
76265-766
EZClick™ Global Protein Synthesis Assay Kit (FACS/Microscopy), Green Fluorescence, BioVision Inc.
Assays Protein Assays
EZClick™ Protein Synthesis Monitoring Assay Kit utilizes a novel and robust chemical method based on an alkyne containing and cell-permeable analog of puromycin, O-propargyl-puromycin (OP-puro).
  • Click-chemistry based assay for measuring de novo protein synthesis in biological samples
  • Simple, fast, does not require lengthy incubation times
Cells generate a complete set of proteins during division. Protein synthesis is a tightly regulated process and many critical controls in gene expression occur at the level of translation to ensure that production of specific cellular proteins is quickly turned on/off under specific conditions (heat shock, starvation, etc.).
Protein synthesis is essential in cell growth, proliferation, signaling, differentiation or death, therefore, the identity and amount of the synthesized proteins are critical parameters in determining the physiological state of the cell. Methods enabling detection and characterization of nascent proteins, or changes in spatial and temporal protein expression/degradation patterns during disease, drug treatments or environmental changes are important tools in assessment of cytotoxicity.
Once inside the cell, OP-puro stops translation by forming covalent conjugates with nascent polypeptide chains. Truncated polypeptides are rapidly turned over by the proteasome and can be detected based on a click reaction with the fluorescent azide.
Unlike methionine analogs, OP-puro does not require methionine-free conditions and can be used to label nascent proteins directly in the cell culture. Our kit provides sufficient materials for 100 assays to detect nascent proteins synthesized under various physiological conditions, and Cycloheximide, an inhibitor of protein synthesis that serves as an experimental control.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty