Would you like to log in again?
PicoProbe™ Threonine Assay Kit (Fluorometric), BioVision
Supplier: BioVision
AB239726-100TEST
76265-742EA 1071.46 USD InStock
76265-742
PicoProbe™ Threonine Assay Kit (Fluorometric), BioVision
Assays Protein Assays
Kit for enzymatic detection of threonine in biological samples. Threonine Assay Kit allows for highly sensitive quantification of L-Threonine levels in biological fluids and tissues. The assay is based on the selective, NAD+-coupled enzymatic metabolism of threonine, yielding an oxidized intermediate and NADH. A developer enzyme mixture utilizes the NADH generated to convert PicoProbe™ into a stable fluorophore.
  • Simple, rapid and convenient assay
  • Limit of detection: 2 µM
  • Very specific, does not interact with other standard and non-standard amino acids
Threonine is one of the 20 naturally-occuring amino acids used by all organisms in the biosynthesis of proteins and is an essential dietary amino acid for humans. Threonine residues in proteins are subject to post-translational phosphorylation, a critical regulatory and signal transduction mechanism.
In addition to its role as a protein building block, threonine can serve as a precursor to other amino acids and metabolites, including glycine, isoleucine and acetyl-coenzyme A. Recent research has shown that threonine is a vital nutrient for embryonic stem cells and threonine metabolism (mediated by the enzyme threonine dehydrogenase) is required for unlimited self-renewal and maintenance of stem cell pluripotency.
Dietary threonine deficiency, while rare, can result in breakdown of the intestinal mucosal lining (eventually triggering inflammatory bowel disorders), immunosuppression and a multitude of neuropsychiatric symptoms.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty