Would you like to log in again?
EZCell™ Intracellular Zinc Staining Kit (Cell-Based), BioVision
Supplier: BioVision
AB241014-50TEST
76265-768EA 959.34 USD InStock
76265-768
EZCell™ Intracellular Zinc Staining Kit (Cell-Based), BioVision
Stains In vivo Imaging Dyes
Flurometric assay for quantitating zinc in a variety of biological samples.
  • Simple procedure takes 2 hours
  • Fast and convenient
  • The assay is sensitive, stable, specific and high-throughput adaptable
  • Visualize intracellular content in suspension and/or adherent cells
Zinc (Zn, Atomic Number: 30) is an essential mineral of great biological significance. It is a metal with characteristics like magnesium due to its size and +2 oxidation states. It is the second most abundant transition metal in living organisms after iron.
Zn2+ binding proteins account for nearly 50% of the transcription regulatory proteins in the human genome and require zinc as an essential cofactor in the regulation of gene expression. Numerous enzymes also employ Zn2+ ions in their active sites. Free Zn2+ is released from metalloprotein complexes during oxidative stress with the remainder being bound to proteins or nucleic acids.
Other examples of biological roles of Zinc include signal transduction, regulation of apoptosis, neurotransmission, synaptic plasticity and prostate gland function. In humans, Zinc deficiency is associated with many chronic diseases such as renal disease, sickle cell anemia, cirrhosis, some cancers and deficiencies in metabolism.
EZCell™ Intracellular Zinc Staining Kit (cell-based) which contains a selective and membrane permeable fluorescent probe for detection of intracellular Zinc. We also include a Positive Control that elevates Zinc staining, and cell-permeable Zinc Chelator that reduces fluorescence induced by Positive Control. This easy-to-use non-radioactive assay allows studies of the regulation of zinc at the cellular level by fluorescence microscopy and flow cytometry in cultured cells.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty