Would you like to log in again?
Soluble Collagen Assay Kit (Fluorometric), BioVision
Supplier: BioVision
AB241015-100TEST
76265-772EA 1054.51 USD InStock
76265-772
Soluble Collagen Assay Kit (Fluorometric), BioVision
Assays Protein Assays
A simple, sensitive and easy fluorometric assay to quantitate soluble collagen in biological samples.
  • Simple, fast and convenient assay
  • Detect as low as 0.05 µg of soluble collagen in a variety of samples
  • Simple procedure, takes less than 60 minutes
  • PCA-free
Collagen is the most abundant protein in mammals, accounting for nearly 30% of body protein content. Collagen is an extracellular scaffolding protein found in skin, tendons, bone, cartilage, ligaments and muscles, amongst other tissues. There are more than twenty different characterized types of collagen, but the fibrilous types I, II and III comprise more than 90% of the total collagen in mammals.
Collagen has a triple helical structure, consisting of three ‘preprocollagen’ strands with a repeating glycine- and proline-rich tripeptide sequence (Gly-Pro-X) and small end-cap domains called registration peptides. Nascent preprocollagen strands undergo enzymatic post-translational hydroxylation at proline and lysine residues prior to secretion to the extracellular milieu via the golgi apparatus.
Once outside the cell, the dangling registration peptides are cleaved off, forming tropocollagen, the monomeric precursor to mature collagen fibers. While mature collagen exists as an insoluble web of extensively crosslinked fibrils, tropocollagen is soluble in dilute acidic solutions. Many diseases are believed to affect collagen synthesis, fibrilization and turnover, including tumor invasion and metastasis, rheumatoid arthritis, cardiopulmonary fibrosis and muscular dystrophy.
Soluble Collagen Assay Kit allows for quantification of newly synthesized (acid-soluble) collagen levels in tissues and cultured cell medium. The assay involves extraction of soluble collagen in acetic acid, followed by enzymatic degradation of collagen into glycine-rich oligopeptides, which are quantified using a fluorogenic reagent and developer solution that selectively react with the N-terminal glycine fragments to form a stable fluorescent complex.
The assay is more sensitive and selective than dye-binding (Picrosirius Red) soluble collagen assays, is simple to perform and has a linear range from 0.05 – 2 μg collagen per well (2.5 μg/ml to 100 μg/ml in a 20 μl sample volume).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty