Would you like to log in again?
D-Amino Acid Oxidase Activity Assay Kit (Fluorometric), BioVision
Supplier: BioVision
AB273325-100TEST
76265-750EA 903.98 USD InStock
76265-750
D-Amino Acid Oxidase Activity Assay Kit (Fluorometric), BioVision
Assays Enzyme Assays
This simple, rapid and convenient assay to measure D-Amino Acid Oxidase activity.
  • The assay can detect as little as 0.5 µU of DAAO enzymatic activity
  • Includes active DAAO as positive control
D-amino acid oxidase (DAAO, DAO, EC 1.4.3.3) is a flavoprotein which oxidizes a D-amino acid forming hydrogen peroxide and the respective imino acid which in a second catalytic step by he same enzyme is hydrolyzed to a α-keto acid and ammonia. In recent years, DAAO has been studied for its possible role in schizophrenia.
The simultaneous expression of DAAO and a putative product from G72 gene may potentially increase the risk of schizophrenia in patients. For example, increased DAAO expression and enzymatic activity have been reported in post mortem brain tissue samples from patients with schizophrenia compared to healthy controls.
DAAO expression in human brain is very specific, it can only be found in the cerebellum and is not detectable in any other parts of the brain. DAAO regulates the physiological levels of D-amino acids such as D-serine, D-aspartate and D-alanine. D-serine acts as a modulator of N-Methyl-D-aspartate receptor (NMDA receptor, NMDAR), while D-aspartate and D-alanine are found to be elevated in both white matter and gray matter in patients’ brains suffering from Alzheimer disease.
D-amino Acid Oxidase Activity Assay Kit provides a quick and easy method for monitoring DAAO activities in a wide variety of samples. In this assay, DAAO converts D-amino acid into hydrogen peroxide as one byproduct. Consequently, the formed byproduct will react with a probe producing a strong fluorescent signal.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty