Would you like to log in again?
PicoProbe ™ Phosphatidic Acid Assay Kit (Fluorometric), BioVision Inc.
Supplier: BioVision
AB273335-100TEST
76265-760EA 1090.1 USD InStock
76265-760
PicoProbe ™ Phosphatidic Acid Assay Kit (Fluorometric), BioVision Inc.
Assays Cellular Assays
The easiest, specific assay kit to measure cardiolipin in biological samples.
  • Simple, fast, does not require lengthy incubation times
  • Detect as low as 40 pmol PA per well
Phospholipids are amphiphilic molecules containing phosphate groups and are both critical components of biological membranes and important signaling messengers in several biological pathways.
Phosphatidic acid (PA) is the simplest phospholipid, consisting of a glycerol backbone with fatty acids bonded to carbon 1 and 2, and a phosphate group bonded to carbon 3. PA is a precursor for the synthesis of more complex phospholipids such as phosphatidylcholine, phosphatidylethanolamine and phosphatidylserine that act as structural components of the plasma membrane.
Phosphatidic acid itself is present in the plasma membrane lending the membrane a high curvature and a net negative charge. In addition, it is an important signaling lipid. It acts through a G-protein coupled receptor and is involved in growth regulation and mitosis.
Phosphatidic acid assay kit is a plate-based enzymatic assay for quantitation of PA in cells and tissues. The PA Converter hydrolyzes PA to form an intermediate which, in the presence of a developer and enzyme mix, converts a non-fluorescent probe to a fluorescent product that can be quantified. The Converter is specific to PA and does not hydrolyze other phospholipids, those with more complex head groups such as phosphatidylcholine or cardiolipin), allowing direct quantitation of PA.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty