Would you like to log in again?
Nitrite Assay Kit (Griess Reagent), BioVision
Supplier: BioVision
AB234044-1000TEST AB234044-200TEST AB234044-500TEST
76265-790EA 1179.77 USD InStock
76265-790 76265-754 76265-756
Nitrite Assay Kit (Griess Reagent), BioVision
Assays Cellular Assays
Nitrite Assay Kit using Griess Reagent provides an accurate, convenient measure of total nitrite. This is an accurate and convenient colorimetric assay to measure nitric oxide from urine, serum.
  • Simple two-step procedure takes only 1 to 1.5 hours
  • Fast and convenient
Nitrogen-based ions, nitrite and nitrate, are found in almost every living organism. Furthermore, endogenous Nitrite levels are found in mammals and can be obtained from dietary sources. In humans, nitrite is further metabolized to Nitric Oxide and other reactive nitrogen species (nitrogen oxides).
The nitrate-nitrite-NO biochemical pathway is well known for its participation in cell signaling, hypoxia-dependent response and regulation of blood flow. Recent studies suggest the reduction of nitrite to nitrogen oxygen in the mitochondria. Specifically, myoglobin and xanthine oxidoreductase could generate NO under hypoxic conditions leading to mitochondrial respiration.
Nitrite Assay Kit utilizes the Griess Reagent, a classic protocol for the estimation of nitrite. In the assay, nitrite is reduced to Nitrogen Oxide using Griess Reagent I. Then, Nitrogen Oxide reacts with Griess Reagent II forming a stable product that can be detected by its absorbance at OD 540 nm. The one-step assay is simple, fast and can detect nitrite levels as low as 1 nmol/well.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty