Would you like to log in again?
Go Direct™ Middle School Packages
Supplier: VERNIER SOFTWARE & TECH SE
GDP-MS-DX
470305-414EA 1117.99 USD InStock
470305-414
Go Direct™ Middle School Packages
Data Loggers
We offer different packages to meet your needs and budget for the middle school classroom.
  • Starter package includes motion detector, pH, voltage, (2) temperature, llght and color probes
  • Deluxe package includes (in addition to starter package) force and acceleration; gas pressure; heart rate; conductivity; 3-axis magnetic field probes
The starter package is great way to get started, and the deluxe package includes all the sensors needed for the experiments in the Middle School Science with Vernier Book item 470017-668.
Our Go Direct™ Probes connects directly to your mobile device, Chromebook, or computer using our free Graphical Analysis™ 4 app-no additional equipment or software purchases are necessary. Go Direct sensors can be used wired via USB or wirelessly via Bluetooth, allowing you to choose the solution best for your classroom or laboratory.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty