Would you like to log in again?
Go Direct™ O₂ Gas Sensor
Supplier: VERNIER SOFTWARE & TECH SE
GDX-O2
470303-910EA 208.99 USD InStock
470303-910
Go Direct™ O₂ Gas Sensor
Data Loggers
The Go Direct O₂ Gas Sensor measures gaseous oxygen concentration levels and air temperature.
  • Test catalase activity under various conditions
  • Measure oxygen consumption at rest and after exercise
  • Measure the change in gas produced during photosynthesis
  • Compare the rates of cell respiration in germinating and non-germinating peas
The Go Direct O₂ Gas Sensor measures gaseous oxygen concentration and air temperature. This sensor has a wide measurement range, which is ideal for studying human and cellular respiration. A 250 ml Nalgene bottle is included for running controlled experiments with small plants and animals.
The Go Direct robust rechargeable battery offers long battery life and provides always-ready operation. Monitor battery life directly from our free Graphical Analysis™ 4 app. Should the battery run low, simply connect the sensor to the charging cable and keep on collecting-no interruptions or inconvenience to you or your students.
Our Go Direct sensors connect directly to your mobile device, Chromebook™, or computer using our free Graphical Analysis™ 4 app-no additional equipment or software purchases are necessary. Go Direct sensors can be used wired via USB or wirelessly via Bluetooth wireless technology, allowing you to choose the solution best for your classroom or laboratory.
Delivery information: Includes Go Direct® O₂ gas sensor, micro USB cable, 250 ml Nalgene bottle with lid.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty