Would you like to log in again?
Go Direct™ Wide Range Temperature
Supplier: VERNIER SOFTWARE & TECH SE
GDX-WRT
470303-848EA 138.99 USD InStock
470303-848
Go Direct™ Wide Range Temperature
Data Loggers
Measuring the temperature of a distillation has never been safer-your students can conduct an experiment within the laboratory fume hood while collecting data at their lab station.
  • Range of –20 to 330 °C allows for investigations such as determining the melting point of caffeine or the boiling point of different vegetable oils
  • RTD (Resistance Temperature Detector) technology establishes a ±0.5 °C accuracy
Designed to be used as you would use a thermometer for experiments such as the recrystallization of benzoic acid, simple and fractional distillations, determination of boiling points, the synthesis and analysis of aspirin and other organic compounds, and more.
The Go Direct robust rechargeable battery offers long battery life and provides always-ready operation. Monitor battery life directly from our free Graphical Analysis™ 4 app. Should the battery run low, simply connect the sensor to the charging cable and keep on collecting-no interruptions or inconvenience to you or your students.
Our Go Direct sensors connect directly to your mobile device, Chromebook™, or computer using our free Graphical Analysis™ 4 app-no additional equipment or software purchases are necessary. Go Direct sensors can be used wired via USB or wirelessly via Bluetooth wireless technology, allowing you to choose the solution best for your classroom or laboratory.
Delivery information: Includes Go Direct wide range temperature, micro USB cable.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty