Would you like to log in again?
Fiber-optic Macintosh Laryngoscope Blade, Flex Style, Sklar®
Supplier: Sklar
07-1449
76191-506EA 734.83 USD InStock
76191-506
Fiber-optic Macintosh Laryngoscope Blade, Flex Style, Sklar®
Scalpels and Scalpel Assemblies Scalpel Blades
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
The Flex-Tip Fiber-Optic Macintosh Laryngoscope Blade, also known as the "English Channel" style blade, is particularly useful during difficult intubations or with patients suffering spinal injuries or displaced larynx, teeth, tongue, or mandible. The Flex-Tip Blade has an attached handle that controls the distal tip of the blade and allows it to move through a 70° angle. This allows the tip to lift the epiglottis and present a clear view of the vocal cords. The Flex-Tip also reduces contact with the teeth and surrounding soft tissue resulting in less trauma for the patient. These fiber-optic blades follow the Green System of fiber-optic compatibility which is indicated by a green bar at the posterior end of the blade where the blade attaches to a fiber-optic handle.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty