Would you like to log in again?
Go Direct™ Chloride Ion Electrode
Supplier: VERNIER SOFTWARE & TECH SE
GDX-CL
470303-902EA 288.99 USD InStock
470303-902
Go Direct™ Chloride Ion Electrode
Electrodes ISE Electrodes
The Go Direct Chloride Ion-Selective Electrode (ISE) is used to measure the concentration of chloride (Cl-) in aqueous samples.
The concentration of chloride gives a quick measurement of the salinity of water samples. Use the Go Direct Chloride Ion-Selective Electrode to wirelessly measure chloride levels in ocean saltwater or salt in food samples.
The versatile BNC connector allows students to easily replace the electrode when it expires or becomes damaged. You need only replace the electrode and not the entire sensor-lowering your overall equipment costs.
The Go Direct Chloride Ion-Selective Electrode has a combination-style, non-refillable, gel-filled electrode. Unlike other ISEs available from Vernier, the Go Direct Chloride Ion-Selective Electrode has a solid-state membrane that does not need to be replaced.
The Go Direct robust rechargeable battery offers long battery life and provides always-ready operation. Monitor battery life directly from our free Graphical Analysis™ 4 app. Should the battery run low, simply connect the sensor to the charging cable and keep on collecting-no interruptions or inconvenience to you or your students.
Our Go Direct sensors connect directly to your mobile device, Chromebook™, or computer using our free Graphical Analysis™ 4 app-no additional equipment or software purchases are necessary. Go Direct sensors can be used wired via USB or wirelessly via Bluetooth wireless technology, allowing you to choose the solution best for your classroom or laboratory.
Certifications: Important: Ion-Selective Electrodes require good chemical technique and careful calibration to obtain accurate results; they are not recommended for middle school or elementary students.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty