Would you like to log in again?
Earth and Human Activity, Water Quality
Supplier: SCIENCE INTERACTIVE GROUP
625-1010
470232-622KT 195 USD InStock
470232-622
Earth and Human Activity, Water Quality
Educational Materials Environmental Educational Materials Environmental Learning Activities
Discover how we impact the quality of our water.
  • Students will explore types of pollution and human impacts to water
  • Modules include in-depth background reading, quizzes, and ideas for lesson extensions that align with NGSS
In the "Earth and Human Activity: Water Quality" kit, students collect water samples from a local environment and test a suite of environmental parameters, including: pH, nitrates, phosphates, oil, and lead. Students are guided through the scientific method and design their own controlled experiments to investigate the effects of nutrient loading on an aquatic system. A teacher demonstration on particle settling time introduces a discussion about particulates in the environment and in drinking water.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty