Would you like to log in again?
BactronEZ Anaerobic Chamber, Compact, 300 Plate Capacity, Sheldon
Supplier: Sheldon
BAAEZ22-2-E BAAEZ22
76097-902EA 20663.21 USD InStock
76097-902 76097-906
BactronEZ Anaerobic Chamber, Compact, 300 Plate Capacity, Sheldon
Anaerobic Systems
BACTRON chambers are total anaerobic process workstations. Samples can be prepared, cultured, and inspected in the oxygen-free chamber and standard incubator. Unlike anaerobic jars and bags, which unavoidably expose samples to oxygen, BACTRON units provide complete anaerobic environments for all your applications.
  • Ergonomic arm ports provide a comfortable work space
  • Integraded vacuum pump For quieter operation
  • 60% faster airlock cycle with 3,4 or 5 auto cycle iterations
  • Auto-sleeve cycle with integrated HEPA filter, 40% faster sleeve cycle
  • Low gas supply audible and visual alarm
  • Single button auto-commissioning
The BactronEZ is an ideal first anaerobic chamber or replacement chamber for users who desire to upgrade their existing chamber. The BactronEZ has a 13.7 cu.ft. workspace and a 300 plate capacity incubator and airlock capacity of 78 Plates. Specifically designed for use by smaller laboratories, it may also serve as an economical set-up for larger labs. Bactron systems have airtight construction of stainless steel and rigid Plexiglas, for unobstructed vision and integrity. In addition, the BactronEZ has our new ergonomic armport doors, which ensure user comfort and minimize shoulder fatigue. The patented cuffs form a comfortable seal around the operator’s arms permitting bare hand manipulation of plates and specimens inside the working chamber, making work more comfortable and efficient.
Chamber atmosphere circulates through a condensate controller then through a catalyst, which removes any trace amounts of oxygen. High relative humidity is maintained while condensate moisture is removed without the unnecessary desiccants, which can dehydrate media and samples. Anaerobes that are exposed to oxygen die or grow more slowly. Unlike anaerobic jars, which expose samples to oxygen during plating, isolation, sub-culturing and susceptibility testing, the BactronEZ is a complete workstation where samples can be prepared, incubated and evaluated in an oxygen free environment. The user has complete access to individual samples within the incubator and may determine the presence of anaerobic bacteria at any time during incubation. In addition, increasing attention to hospital acquired infections has made quick, accurate detection of anaerobes that cause infections extremely important.
Unlike many anaerobic chamber manufacturers, the SHEL LAB Bactron chambers have full third party verified safety compliance certificates and meet EN61010, UL61010 and Can/CSA61010 standards.
See also our BACTRON300, BACTRON600 AND BACTRON900 models.
Certifications: CE/TUV, EN61010, CAN/CSA61010.
Ordering information: 1-year limited warranty.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty