Would you like to log in again?
Richter Dissecting Scissors, OR-Grade, Sklar®
Supplier: Sklar
15-1276
76193-052EA 308.42 USD InStock
76193-052
Richter Dissecting Scissors, OR-Grade, Sklar®
Scissors
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
Richter Dissecting Scissors are used primarily for separating tissue or cutting specimen for study. These standard scissors should be used with tissue that is neither too delicate nor too tough--cases where Mayo or Metzenbaum dissecting scissors would be more appropriate.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty