Would you like to log in again?
Sklarlite™ Metzenbaum-Nelson Bariatric Dissecting Scissors, OR-Grade, Sklar®
Supplier: Sklar
15-3309 15-3319
76191-744EA 369.76 USD InStock
76191-744 76191-746
Sklarlite™ Metzenbaum-Nelson Bariatric Dissecting Scissors, OR-Grade, Sklar®
Scissors
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
Supercut SklarHone™ SklarLite™ Metzenbaum-Nelson Bariatric Scissors are long dissecting scissors most commonly used in Bariatric Surgeries such as gastric bypass, gastric banding, and sleeve gastrectomy. They are also used in Nephrectomy, Cystectomy, and Prostatectomy procedures to dissect and cut tissue. Supercut Sklarhone™ Sklarlite™ scissors provide lightweight, precise cutting. These scissors feature one knife-like razor edge and one micro-serrated edge combined with a unique manufacturing process that shaves microns off instruments while maintaining structural integrity and durability. This lightweight modification provides the surgeon with greater control and less fatigue. Supercut Sklarhone™ Sklarlite™ instruments are identified by dual black finger rings.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty