Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
RNase R
Supplier: BioVision
AB286929-1KIT
76265-830EA 599.72 USD InStock
76265-830
RNase R
Enzymes
An E. coli exoribonuclease
  • Simple procedure
  • Fast and convenient
RNase R is an E. coli exoribonuclease which exhibits 3’-to-5’ exonuclease activity, efficiently digesting nearly all linear RNA species.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty