Would you like to log in again?
ToxOut™ Rapid Removal Wash Buffer , BioVision Inc.
Supplier: BioVision
AB278801-50ML
76265-684EA 268.11 USD InStock
76265-684
ToxOut™ Rapid Removal Wash Buffer , BioVision Inc.
Buffers Biological Buffers
50 ml of Wash buffer for ToxOut Endotoxin Removal Agarose after the regeneration procedure
  • Simple procedure
  • Fast, effective and convenient
Endotoxin is the lipopolysaccharide (LPS) complex located in the outer membrane of gram-negative bacteria. During experimental procedures, large amount of endotoxins are shed and can easily contaminate labware, buffers and downstream products. Therefore, it is critical to remove endotoxin from samples and products. ToxOutTM Rapid Removal Wash Buffer is the endotoxin-free wash buffer containing high concentration of salt for washing BioVision’s ToxOutTM Rapid Endotoxin Removal Agarose after the regeneration procedure. ToxOutTM Rapid Endotoxin Removal Agarose can effectively reduce endotoxins to < 0.05 EU/mL in solutions containing proteins or pharmacologically important components.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty