Would you like to log in again?
ExoPure™ Maxi Chromatography Columns, BioVision, Inc.
Supplier: BioVision
AB288124-3UNITS AB288124-6UNITS
76200-134EA 2029 USD InStock
76200-134 76200-136
ExoPure™ Maxi Chromatography Columns, BioVision, Inc.
Chromatography Columns
ExoPure™ Chromatography Columns is a Size Exclusion Chromatography (SEC) column designed for isolating highly pure exosomes from urine and cell media.
  • Easy and fast protocol (approximately 15 minutes)
  • Isolate exosome from diluted matrices and different sample types
  • Easy to ship and store at 4 °C
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty