Would you like to log in again?
MTS Cell Proliferation Colorimetric Assay Kit, BioVision
Supplier: BioVision
AB197010-10000TEST AB197010-250TEST AB197010-5000TEST
76200-042EA 1454.91 USD InStock
76200-042 76200-044 76200-040
MTS Cell Proliferation Colorimetric Assay Kit, BioVision
Assays Cellular Assays
Faster and sensitive MTS based assay to measure cell proliferation.
  • Simple one-step procedure, takes approximately 4 hours
  • High-throughput
  • The entire assay can be performed directly in a 96-well plate and does not require washing/harvesting/or solubilization steps
MTS cell proliferation assay kit is a colorimetric method for sensitive quantification of viable cells in proliferation and cytotoxicity assay. The method is based on the reduction of MTS tetrazolium compound by viable cells to generate a colored formazan product that is soluble in cell culture media.
This conversion is thought to be carried out by NAD(P)H-dependent dehydrogenase enzymes in metabolically active cells. The formazan dye produced by viable cells can be quantified by measuring the absorbance at 490 to 500 nm. The assay can be used for the measurement of cell proliferation in response to growth factors, cytokines, mitogens, and nutrients and more.
It can also be used for the analysis of cytotoxic compounds like anticancer drugs and many other toxic agents and pharmaceutical compounds. It is performed by adding the reagent directly into the cell culture media without the intermittent steps, which are required in the routine MTT assay.
In addition, this high-throughput assay requires no washing or solubilization step and can be performed in 96-well microtiter plate.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty