Would you like to log in again?
Lipopolysaccharide (LPS) Isolation Kit, BioVision
Supplier: BioVision
AB239718-10TEST
76200-102EA 408.67 USD InStock
76200-102
Lipopolysaccharide (LPS) Isolation Kit, BioVision
Assays Cellular Assays
Simple, one step assay to isolate PBMC.
  • This kit does not use chlorofrom or phenol
  • Yields pure LPS in less than 2 hours that can be easily characterized and quantified
The outer membrane of gram-negative bacteria contains lipopolysaccharide (LPS), a low molecular weight carbohydrate with a molecular mass of 10 to 20 kDa. It is heterogeneous, and composed of O antigen (a repeating glycan polymer), core oligosaccharide (which links the O antigen to Lipid A - the third component, and non-carbohydrate components such as phosphate and amino acids groups.
Lipid A, has multiple fatty acids which serve to anchor LPS into the bacterial membrane allowing the O antigen and core oligosaccharide to protrude and contributes to a major part of the toxicity of gram-negative bacteria. Also known as endotoxin, when consumed by animals, LPS induces a strong inflammatory response and/or sepsis.
The LPS Isolation Kit uses bacterial membrane lysis buffer and protein digestion to yield micrograms of LPS from bacterial culture (approximately 1 to 4% of dry weight).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty