Would you like to log in again?
Human Whole Blood Polymorphonuclear Cell Isolation Kit, BioVision
Supplier: BioVision
AB241001-30TEST
76200-056EA 580.73 USD InStock
76200-056
Human Whole Blood Polymorphonuclear Cell Isolation Kit, BioVision
Assays Cellular Assays
Simple, one step assay to isolate PMN.
  • PMN high yields (≥5×106 cells/ml of human whole bloodl)
  • By using the viability stain, it was proven that 99% of the cells are viable and the isolated fraction contains low red blood cells (≤ 3%)
Polymorphonuclear cells (PMN) have a multilobed nucleus and include the following cells types: mast cells, neutrophils, eosinophils and basophils. Upon isolation, these cells can be used for migration assays and determination of cytokine/chemokine production in response to pharmacological compounds.
PMN cell isolation kit is unique in the market because it enables the user to isolate PMNs, assess viability of the obtained cell fractions and also determine purity with three simple, easy-to-use reagents.
Purity of the cell fraction is assessed with giemsa stain. The kit uses results in highly enriched (>95%) cell fraction of polymorphonuclear cells with less than 3% red blood cell counts. More than 99% of the cells are viable, as determined by the viability stain.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty