Would you like to log in again?
EZClick™ Global Phospholipid Synthesis Assay Kit (FACS/Microscopy), Red Fluorescence, BioVision Inc.
Supplier: BioVision
AB241025-100TEST
76200-048EA 903.8 USD InStock
76200-048
EZClick™ Global Phospholipid Synthesis Assay Kit (FACS/Microscopy), Red Fluorescence, BioVision Inc.
Assays Cellular Assays
Chemistry based assay for measuring de novo phospholipid synthesis in biological samples.
  • Simple, fast, does not require lengthy incubation times
Phospholipids are major component of the bilayers of all plasma membranes. A single phospholipid molecule consists of a phosphate group on one end, called the "head", and two side-by-side chains of fatty acids that make up the "tails". The phosphate head groups can be modified with organic molecules such as Choline (Cho). Cho-containing phospholipids (Phosphatidylcholines - PC) are critical for structural membrane integrity, cellular metabolism and signaling either as individual molecules or precursors of secondary messengers.
Changes in global synthesis of Cho-containing phospholipids are an essential parameter in analysis of cellular response to both, physiological and pathological conditions, environmental stress, or drug treatment. To date, phospholipids biochemistry, cell biology and metabolism remain obscure, due to limited methods for their direct cellular visualization.
EZClick™ Global Phospholipid Synthesis Assay Kit offers a simple and robust method to label and visualize newly synthesized phospholipids in vivo. Based on the metabolic incorporation of the choline analogs directly into their structure, modified phospholipid molecules can be detected with high sensitivity and spatial resolution by click chemistry with azide-containing dyes.
This kit enables analyses of global biosynthesis, subcellular localization and turnover of Cho-containing phospholipids in cells. Cells show strong incorporation of Cho analogs into all classes of phospholipids that can be assayed by fluorescence microscopy, or quantified by FACS. We provide sufficient materials for 100 assays.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty