Would you like to log in again?
CENCO® Hall Effect
Supplier: VWR International
Ward's®
HLEF01-VWR
470303-942EA 3017.05 USD InStock
470303-942
CENCO® Hall Effect
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Explore the Hall Effect in a GaAs semiconductor.
  • n-type GaAs Hall sensor chip
  • Helmholtz coil pair 75 mm effective diameter, 11.25 mT central magnetic induction at 0.5 A
  • Two-axis mechanical traverse for field plotting
  • Constant current sources for magnet (0 to 0.5 A) and Hall driving current (0 to 3 mA)
  • Reversing switch for exploring the elimination of systematic error
  • 31/2 digit current and voltage meters
  • Metal storage case for the Hall Effect board
Explores systematic errors and their elimination. Determines the conductivity of the semiconductor.
This Hall Effect Apparatus consists of a mounted n-type semiconductor chip on a traverse mechanism, a pair of coils, relay-controlled reversing switches, and a control unit with connecting cords.
Uses the semiconductor to plot the coils magnetic field.
The Hall Effect is the generation of a side-to-side voltage in a conductor or semiconductor carrying a current when it is placed in a magnetic field.
The effect is widely used in magnetic field sensors. It is associated with several well-known systematic errors, some of which can be eliminated by special measurement techniques.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty