Would you like to log in again?
Heating Mantles
Supplier: United Scientific Supplies
Ward's®
HM1000-250 HM1000-1000 HM1000-100 HM1000-500
470303-950EA 300.08 USD InStock
470303-950 470303-944 470303-946 470303-948
Heating Mantles
Heaters Heating Mantles
Designed for use with round bottom flasks.
  • 4' cord
  • All steel construction
Mantles can be used in a wide range of ambient temperatures, from about 45 to 100 °F, and relative humidity up to 80%.
A 4-foot long grounded cord is included.
Features include polyurethane coated aluminum exteriors and a high quality knitted mantle with glass fiber underlay for uniform distribution of heat. Maximum operating temperature is 400°C.The units are not waterproof, and are rated as IP code 10 equipment.
Power is controlled (from 0 to 100%) by a knob that is built into the unit - no need for a separate external power control unit. Mantles are designed to securely hold flasks so that the top half of the flask is easily visible during operation. Elements can be replaced easily. An outlet tube is built-in for draining any spilled liquids. Each unit features a fuse that can be easily replaced.
Units are designed for long-term use. Care should be taken to avoid chemical spillage, overheating, overloading, and general misuse to maximize service life. Please refer to the user manual for more information.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty