Would you like to log in again?
Activated Protein C (APC) Inhibitor Screening Kit (Colorimetric), BioVision, Inc.
Supplier: BioVision
AB283383-100TEST
76200-070EA 1285.66 USD InStock
76200-070
Activated Protein C (APC) Inhibitor Screening Kit (Colorimetric), BioVision, Inc.
Assays Protein Assays
Quick and easy screening for activated protein C ligands.
  • Simple, highly sensitive
  • Rapid screening and high-throughput compatible
Activated protein C (APC) (3.4.21.69), a Vitamin K dependent serine protease, is formed by activation of protein C by thrombomodulin bound on the surface of endothelial cells. By degrading coagulation factors Va and VIIIa, APC inhibits blood coagulation which helps in the prevention of thrombosis.
Blood coagulation can be stimulated by inhibition of APC. Thus, APC inhibitors play an important role in the treatment of disease like haemophilia.This Kit can be used to screen for potent inhibitors of APC activity. It utilizes the proteolytic activity of an active APC on a peptide substrate which releases a colored product.
The released product can be easily quantified using an absorbance microplate reader. In the presence of an APC inhibitor, the enzyme decreases/loses activity resulting in lower/no absorbance. This assay kit is simple and can be used to identify and characterize APC inhibitors in a high-throughput format. As a control, kit is provided with an inhibitor PPACK dihydrochloride - a known APC inhibitor.
Delivery information: Kit includes PPACK (APC inhibitor) and a stable, recombinant human APC
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty