Would you like to log in again?
Human Inosine-5’-Monophosphate Dehydrogenase (IMPDH) Inhibitor Screening Assay Kit, BioVision, Inc.
Supplier: BioVision
AB283395-100TEST
76200-066EA 903.37 USD InStock
76200-066
Human Inosine-5’-Monophosphate Dehydrogenase (IMPDH) Inhibitor Screening Assay Kit, BioVision, Inc.
Assays Enzyme Assays
High-throughput compatible, quick and easy screening for neprilysin inhibitors.
  • Simple and reliable test to screen IMPDH inhibitors/activators
  • High-throughput compatible
  • With inhibitor control (MPA)
Inosine monophosphate dehydrogenase, IMPDH, (E.C. 1.1.1.205) is the rate-limiting enzyme in de novo guanine nucleotide biosynthesis and it is essential for lymphocyte proliferation. IMPDH oxidizes inosine 5’-monophosphate (IMP) to xanthine 5’-monophosphate (XMP) using NAD as a cofactor. It plays a critical role in cell growth and in the malignancy of some tumors such as pancreatic, colon and bladder cancers.
IMPDH2 is the predominant isoform of IMPDH and is specifically linked to a wide range of cancers and lymphocyte proliferation. It is recognized as a validated target as antiviral, antiparasitic, antimicrobial, antileukemic and immunosuppressive agents. In this inhibitor's screening assay, IMP is oxidized by IMPDH producing a series of intermediates, which react with the probe to generate a colorimetric signal (OD 450 nm).
The signal is directly proportional to the IMPDH activity and has a very low background. In the presence of a IMPDH inhibitor such as mycophenolic acid (MPA), the reaction is arrested, thus decreasing the signal. The assay is fast, sensitive and reproducible and is suitable for functional assays, high-throughput screening and preclinical studies in drug discovery.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty