Would you like to log in again?
PicoProbe™ 15-PGDH Activity Assay Kit (Fluorometric), BioVision, Inc.
Supplier: BioVision
AB273327-100TEST
76200-068EA 967.97 USD InStock
76200-068
PicoProbe™ 15-PGDH Activity Assay Kit (Fluorometric), BioVision, Inc.
Assays Enzyme Assays
Highly sensitive fluorometric assay for measuring PGDH activity in various biological samples.
  • Rapid, simple, sensitive, and reliable test procedure
  • Fast and convenient
  • Suitable for detecting as low as 1 µU of 15-PGDH activity
15-Hydroxy prostaglandin dehydrogenase (15-PGDH; EC:1.1.1.141) is a rate-limiting enzyme for prostaglandin catabolism. It oxidizes prostaglandins (PG) to 15-keto metabolites. In general, 15-keto metabolites show reduced biological activity, when compared to PG.
PG's are generated from arachidonic acid by cyclooxygenase (COX), and play an important role in inflammatory responses. Recent study found that 15-PGDH serves as a degrading enzyme for PG and serves as a tumor suppressor to inhibit cancer progression. Thus, study of 15-PGDH activity can elucidate and clarify unknown aspects related to cancer development and therapeutics.
The assay Kit, 15-PGDH oxidizes a substrate forming intermediates and NADH. The oxidation of NADH reduces a probe generating fluorescence at Ex/Em = 535/587 nm. The activity of 15-PGDH is proportional to the fluorescent signal.
Sensitive assay for measuring 15-PGDH activity in various biological samples and the kit is also suitable for high throughput analyses.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty