Would you like to log in again?
Fiber-optic Miller Laryngoscope Blade, English Style, Sklar®
Supplier: Sklar
07-1462
76193-096EA 239.85 USD InStock
76193-096
Fiber-optic Miller Laryngoscope Blade, English Style, Sklar®
Forceps
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
Miller Laryngoscope Blades are used for direct, rigid laryngoscopy procedures to view vocal cords, remove foreign objects from the throat, biopsy, or tracheal intubation. The Miller blade is straight and therefore longer than the curved Macintosh blade. It is inserted past the epiglottis, typically causing less trauma during intubation. The Miller blade is commonly used with pediatric and infant patients. This product is a standard, 3 mm, #00 American Style Miller blade. It has a working length of 40 mm and an overall length of 76 mm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty