Would you like to log in again?
Proteinase K
Supplier: Agilent Technologies
300140 300140
ST300140EA 184.04 USD InStock
ST300140 CAST300140
Proteinase K
Enzymes
Proteinase K is a broad spectrum protease from Tritirachium album that is ideal for preparing chromosomal DNA.
  • For general digestion of protein in biological samples
  • Lyophilized powder with specific activity >30 units/mg at 37 °C
It cleaves peptide bonds in which carbonyl groups are surrounded by aromatic, hydrophobic, or aliphatic amino acids.
Used for the following applications: pulsed field electrophoresis, chromosomal DNA for protein fingerprinting, and nuclease removal from DNA and RNA preparations.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty